• USA Home
  • Search Results

  • neocuproinehydrochloridehydrate24472anhydrousbasis33236000611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Neocuproine hydrochloride hydrate

1 Products
Synonym:   2,9-Dimethyl-1,10-phenanthroline hydrochloride hydrate
Image of Neocuproine hydrochloride hydrate
CAS Number:
Empirical Formula (Hill Notation):
 C14H12N2 · HCl · xH2O
Molecular Weight:
 244.72 (anhydrous basis)
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More