• USA Home
  • Search Results

  • neocuproinehydrochloridemonohydrate2627330313682511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Neocuproine hydrochloride monohydrate

1 Products
Synonym:   2,9-Dimethyl-1,10-phenanthroline hydrochloride
Image of Neocuproine hydrochloride monohydrate
CAS Number:
Empirical Formula (Hill Notation):
 C14H12N2 · HCl · H2O
Molecular Weight:
 262.73
 Beilstein/REAXYS Number:
 4584190
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More