• USA Home
  • Search Results

  • neodiosmin608543866501911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Neodiosmin

1 Products
Synonym:   3′,5,7-Trihydroxy 4′-methoxyflavone 7-neohesperidoside, 7-[[2-O-(6-Deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl]oxy]-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)- 4H-1-benzopyran-4-one, Diosmetin 7-neohesperidoside, Neodiosmin
Image of Neodiosmin
CAS Number:
Empirical Formula (Hill Notation):
 C28H32O15
Molecular Weight:
 608.54
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More