• USA Home
  • Search Results

  • neohesperidin610561324133311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Neohesperidin

4 Products
Synonym:   (S)-4′-Methoxy-3′,5,7-trihydroxyflavanone-7-[2-O-(α-L-rhamnopyranosyl)-β-D-glucopyranoside], Hesperetin 7-O-neohesperidoside, NSC 31048
Image of Neohesperidin
CAS Number:
Empirical Formula (Hill Notation):
 C28H34O15
Molecular Weight:
 610.56
 Beilstein/REAXYS Number:
 74945
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Special Grade
  Application(s)
  Manufacturer designation
Show More