• USA Home
  • Search Results

  • neohesperidindihydrochalcone612582070277611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Neohesperidin dihydrochalcone

5 Products
Synonym:   1-(4-((2-O-[6-Deoxy-α-L-mannopyranosyl]-β-D-glucopyranosyl)oxy)-2,6-dihydroxyphenyl)-3-[3-hydroxy-4-methoxyphenyl]-1-propanone, NHDC
Image of Neohesperidin dihydrochalcone
CAS Number:
Empirical Formula (Hill Notation):
 C28H36O15
Molecular Weight:
 612.58
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Anal. Standard Form
  Physical Form
  Special Grade
  Assay
Show More