• USA Home
  • Search Results

  • neomycinsulphate61464freebasebasis140510311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Neomycin sulphate

1 Products
CAS Number:
Empirical Formula (Hill Notation):
 C23H46N6O13 · xH2SO4
Molecular Weight:
 614.64 (free base basis)
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More