• USA Home
  • Search Results

  • neomycintrisulfatesalthydrate90888anhydrousbasis140510311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Neomycin trisulfate salt hydrate

5 Products
Image of Neomycin trisulfate salt hydrate
CAS Number:
Empirical Formula (Hill Notation):
 C23H46N6O13 · 3H2SO4 · xH2O
Molecular Weight:
 908.88 (anhydrous basis)
 EC Number:
 NACRES:
 NA.76
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More