• USA Home
  • Search Results

  • nervonicacid3666250637611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nervonic acid

2 Products
Synonym:   cis-15-Tetracosenoic acid, Selacholeic acid
Image of Nervonic acid
CAS Number:
Linear Formula:
 CH3(CH2)7CH=CH(CH2)13COOH (cis)
Molecular Weight:
 366.62
 Beilstein/REAXYS Number:
 1728551
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Anal. Standard Form
  Shelf Life
  Special Grade
  Ice Shipping
  Functional group
  Physical Form
  Application(s)
Show More