• USA Home
  • Search Results

  • nethylethylenediamine881511072511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-Ethylethylenediamine

1 Products
Synonym:   2-Ethylaminoethylamine
Image of <I>N</I>-Ethylethylenediamine
CAS Number:
Linear Formula:
 C2H5NHCH2CH2NH2
Molecular Weight:
 88.15
 Beilstein/REAXYS Number:
 1098272
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More