• USA Home
  • Search Results

  • neuraminidasefromclostridiumperfringenscwelchii12345900167611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Neuraminidase from Clostridium perfringens (C. welchii)

4 Products
Synonym:   Acyl-neuraminyl Hydrolase, Receptor-destroying enzyme, Sialidase
Image of Neuraminidase from <I>Clostridium perfringens (C. welchii)</I>
CAS Number:
 EC Number:
 Enzyme Commission (EC) Number:
 3.2.1.18  ( BRENDA | IUBMB )
 MDL number:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Type
  Chemical Composition
  Specific Activity
  Physical Form
Show More