• USA Home
  • Search Results

  • neuraminidasefromvibriocholerae12345900167611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Neuraminidase from Vibrio cholerae

2 Products
Synonym:   Acyl-neuraminyl Hydrolase, Receptor-destroying enzyme, Sialidase
Image of Neuraminidase from <I>Vibrio cholerae</I>
CAS Number:
 EC Number:
 Enzyme Commission (EC) Number:
 3.2.1.18  ( BRENDA | IUBMB )
 MDL number:
 NACRES:
 NA.54
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Type
  Physical Form
Show More