• USA Home
  • Search Results

  • neutralred2887855324211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Neutral Red

5 Products
Synonym:   Basic Red 5
Image of Neutral Red
CAS Number:
Molecular Weight:
 288.78
 Colour Index Number:
 50040
 EC Index Number
 209-035-8
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Chemical Composition
  Special Grade
Show More