• USA Home
  • Search Results

  • nglycolylneuraminicacid32527111383311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-Glycolylneuraminic acid

2 Products
Synonym:   Neu5Glc, NeuNGl
Image of N-Glycolylneuraminic acid
CAS Number:
Empirical Formula (Hill Notation):
 C11H19NO10
Molecular Weight:
 325.27
 Beilstein/REAXYS Number:
 1716828
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More