• USA Home
  • Search Results

  • nheptafluorobutyrylimidazole264103247735311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-Heptafluorobutyrylimidazole

2 Products
Synonym:   1-(Perfluorobutyryl)imidazole, NSC 151966
Image of <I>N</I>-<WBR>Heptafluorobutyrylimidazole
CAS Number:
Empirical Formula (Hill Notation):
 C7H3F7N2O
Molecular Weight:
 264.10
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More