• USA Home
  • Search Results

  • nheptane1002014282511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

n-Heptane

8 Products
Synonym:   Heptane
Image of n-Heptane
CAS Number:
Linear Formula:
 CH3(CH2)5CH3
Molecular Weight:
 100.20
 Beilstein/REAXYS Number:
 1730763
 EC Index Number:
 205-563-8
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Potency
  Application(s)
  Solubility
  Physical Form
  Transition temperature
  Assay
Show More