• USA Home
  • Search Results

  • nhexane861811054311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

n-Hexane

11 Products
Synonym:   n-Hexane, Hexane
Image of n-Hexane
CAS Number:
Linear Formula:
 CH3(CH2)4CH3
Molecular Weight:
 86.18
 Beilstein/REAXYS Number:
 1730733
 EC Index Number:
 203-777-6
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Potency
  Special Grade
  Solubility
  Application(s)
  Physical Form
  Viscosity
  Transition temperature
  Assay
Show More