• USA Home
  • Search Results

  • nickeliichloridehexahydrate23769779120011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nickel(II) chloride hexahydrate

6 Products
Synonym:   Nickel chloride hexahydrate 2, Nickel dichloride hexahydrate
Image of Nickel(II) chloride hexahydrate
CAS Number:
Linear Formula:
 NiCl2 · 6H2O
Molecular Weight:
 237.69
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Restriction
  pH Value
  Product Line
  Reaction Suitability
  Special Grade
  Assay
Show More