• USA Home
  • Search Results

  • nickeliiiodide312501346290311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nickel(II) iodide

2 Products
Synonym:   Nickel diiodide, Nickel iodide, Nickelous iodide
Image of Nickel(II) iodide
CAS Number:
Linear Formula:
 NiI2
Molecular Weight:
 312.50
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Physical Form
Show More