• USA Home
  • Search Results

  • nickeliioxide7469131399111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nickel(II) oxide

5 Products
Synonym:   Nickel monooxide, Nickelous oxide
Image of Nickel(II) oxide
CAS Number:
Linear Formula:
 NiO
Molecular Weight:
 74.69
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Color
  Particle Dimension
  Assay
  Physical Form
Show More