• USA Home
  • Search Results

  • nicotinamide122129892011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nicotinamide

7 Products
Synonym:   Niacinamide, Nicotinic acid amide, Pyridine-3-carboxylic acid amide, Vitamin B3, Vitamin PP
Image of Nicotinamide
CAS Number:
Empirical Formula (Hill Notation):
 C6H6N2O
Molecular Weight:
 122.12
 Beilstein/REAXYS Number:
 383619
 EC Number:
 MDL number:
 NACRES:
 NA.75
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  pH Value
  Transition temperature
  Solubility
  Manufacturer designation
  Density
  Bulk Density
  Autoignition temp
  Special Grade
  Application(s)
  Potency
  Melting Point
  Biological Source
  Agency/Method
  Color
  Assay
Show More