• USA Home
  • Search Results

  • nicotinamideniacinamide122129892011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nicotinamide (Niacinamide)

1 Products
Synonym:   Niacinamide, Nicotinamide, Nicotinic acid amide, Pyridine-3-carboxylic acid amide, Vitamin B3, Vitamin PP
Image of Nicotinamide (Niacinamide)
CAS Number:
Empirical Formula (Hill Notation):
 C6H6N2O
Molecular Weight:
 122.12
 Beilstein/REAXYS Number:
 383619
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More