• USA Home
  • Search Results

  • nicotinicacid123115967611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nicotinic acid

10 Products
Synonym:   Niacin, 3-Pyridinecarboxylic acid, Nicotinic acid
Image of Nicotinic acid
CAS Number:
Linear Formula:
 3-(COOH)-C5H4N
Molecular Weight:
 123.11
 EC Index Number
 200-441-0
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Agency/Method
  Transition temperature
  Solubility
  Purity Grade
  Molecular weight
  Density
  Bulk Density
  Autoignition temp
  Anal. Standard Form
  pH Value
  Potency
  Melting Point
  Titertest
  Biological Source
  Special Grade
  Assay
  Color
Show More