• USA Home
  • Search Results

  • nifedipine346332182925411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nifedipine

5 Products
Synonym:   1,4-Dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylic acid dimethyl ester
Image of Nifedipine
CAS Number:
Empirical Formula (Hill Notation):
 C17H18N2O6
Molecular Weight:
 346.33
 EC Number:
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Titertest
  Solubility
  Anal. Standard Form
  Special Grade
Show More