• USA Home
  • Search Results

  • nifuroxazide2752296552611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nifuroxazide

2 Products
Synonym:   5-Nitro-2-furaldehyde p-hydroxybenzoylhydrazone
Image of Nifuroxazide
CAS Number:
Empirical Formula (Hill Notation):
 C12H9N3O5
Molecular Weight:
 275.22
 Beilstein/REAXYS Number:
 4264976
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Anal. Standard Form
  Physical Form
Show More