• USA Home
  • Search Results

  • nimesulide308315180378211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nimesulide

3 Products
Synonym:   N-(4-Nitro-2-phenoxyphenyl)methanesulfonamide
Image of Nimesulide
CAS Number:
Empirical Formula (Hill Notation):
 C13H12N2O5S
Molecular Weight:
 308.31
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
Show More