• USA Home
  • Search Results

  • niobium9291744003111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Niobium

418 Products
Synonym:   Columbium
Image of Niobium
CAS Number:
Empirical Formula (Hill Notation):
 Nb
Molecular Weight:
 92.91
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Anal. Standard Form
  Special Grade
  Weight
  Package quantity
  Particle Dimension
  Assay
  Physical Form
Show More

Product #
Description
262722 Niobium, powder, <45 μm, 99.8% trace metals basis,
262781 Niobium, foil, thickness 0.25 mm, 99.8% trace metals basis,
IRMM525A Niobium, IRMM® certified Reference Material, 0.02 mm foil,
IRMM525C Niobium, IRMM® certified Reference Material, 0.5 mm wire,
IRMM526A Niobium, IRMM® certified Reference Material, 0.02 mm foil,
IRMM526B Niobium, IRMM® certified Reference Material, 0.1 mm foil,
IRMM526C Niobium, IRMM® certified Reference Material, 0.5 mm wire,
GF76912308 Niobium, insulated wire, 20m, conductor diameter 0.125mm, insulation thickness 0.01mm, polyimide insulation,
GF20291802 Niobium, insulated wire, 2m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyimide insulation,
GF97645090 Niobium, insulated wire, 2m, conductor diameter 0.05mm, insulation thickness 0.008mm, PTFE (polytetrafluoroethylene) insulation,
GF78679333 Niobium, insulated wire, 2m, conductor diameter 0.125mm, insulation thickness 0.01mm, polyimide insulation,
GF36475353 Niobium, insulated wire, 5m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyimide insulation,
GF05636538 Niobium, insulated wire, 5m, conductor diameter 0.05mm, insulation thickness 0.008mm, PTFE (polytetrafluoroethylene) insulation,
GF41596825 Niobium, insulated wire, 5m, conductor diameter 0.125mm, insulation thickness 0.01mm, polyimide insulation,
GF03037115 Niobium, microfoil, disks, 10mm, thinness 0.025μm, specific density 21μg/cm2, permanent mylar 3.5μm support, 99.9%,
GF54977338 Niobium, microfoil, disks, 25mm, thinness 0.025μm, specific density 21μg/cm2, permanent mylar 3.5μm support, 99.9%,
GF01994700 Niobium, pellets, 100g, max. size 10mm, 99.9%,
GF49338120 Niobium, pellets, 200g, max. size 10mm, 99.9%,
GF66415956 Niobium, pellets, 50g, max. size 10mm, 99.9%,
GF76721610 Niobium, rod, 1000mm, diameter 10.0mm, annealed, 99.9%,
GF04176121 Niobium, rod, 1000mm, diameter 2.0mm, annealed, 99.9%,
GF56635948 Niobium, rod, 1000mm, diameter 5.0mm, annealed, 99.9%,
GF61863914 Niobium, rod, 1000mm, diameter 5.6mm, annealed, 99.9%,
GF90201428 Niobium, rod, 100mm, diameter 12.7mm, annealed, 99.9%,
GF23593428 Niobium, rod, 100mm, diameter 2.0mm, annealed, 99.9%,
GF71082031 Niobium, rod, 100mm, diameter 20.0mm, annealed, 99.9%,
GF52663653 Niobium, rod, 100mm, diameter 25mm, 99.9%,
GF36547724 Niobium, rod, 100mm, diameter 30.0mm, annealed, 99.9%,
GF40172600 Niobium, rod, 100mm, diameter 42mm, annealed, 99.9%,
GF24904784 Niobium, rod, 100mm, diameter 5.6mm, annealed, 99.9%,
GF80555317 Niobium, rod, 200mm, diameter 10.0mm, annealed, 99.9%,
GF75178345 Niobium, rod, 200mm, diameter 12.7mm, annealed, 99.9%,
GF49177862 Niobium, rod, 200mm, diameter 2.0mm, annealed, 99.9%,
GF85254396 Niobium, rod, 200mm, diameter 20.0mm, annealed, 99.9%,
GF19789879 Niobium, rod, 200mm, diameter 5.6mm, annealed, 99.9%,
GF95422147 Niobium, rod, 25mm, diameter 20.0mm, annealed, 99.9%,
GF28627106 Niobium, rod, 25mm, diameter 30.0mm, annealed, 99.9%,
GF69413777 Niobium, rod, 500mm, diameter 10.0mm, annealed, 99.9%,
GF97292504 Niobium, rod, 500mm, diameter 12.7mm, annealed, 99.9%,
GF98853081 Niobium, rod, 500mm, diameter 5.0mm, annealed, 99.9%,
GF40950839 Niobium, rod, 500mm, diameter 5.6mm, annealed, 99.9%,
GF51077965 Niobium, rod, 50mm, diameter 12.7mm, annealed, 99.9%,
GF27768651 Niobium, rod, 50mm, diameter 20.0mm, annealed, 99.9%,
GF50557709 Niobium, rod, 50mm, diameter 30.0mm, annealed, 99.9%,
GF00873503 Niobium, rod, 50mm, diameter 42mm, annealed, 99.9%,
GF26609834 Niobium, tube, 1000mm, outside diameter 1.0mm, inside diameter 0.5mm, wall thickness 0.25mm, as drawn, 99.9%,
GF61482539 Niobium, tube, 1000mm, outside diameter 10.0mm, inside diameter 9.2mm, wall thickness 0.4mm, as drawn, 99.9%,
GF14382794 Niobium, tube, 1000mm, outside diameter 2.54mm, inside diameter 2.44mm, wall thickness 0.05mm, annealed, 99.9%,
GF67265039 Niobium, tube, 100mm, outside diameter 1.0mm, inside diameter 0.5mm, wall thickness 0.25mm, as drawn, 99.9%,
GF36644441 Niobium, rod, 100mm, diameter 10.0mm, annealed, 99.9%,
GF22015921 Niobium, rod, 100mm, diameter 5.0mm, annealed, 99.9%,
GF51990824 Niobium, rod, 200mm, diameter 5.0mm, annealed, 99.9%,
GF47233403 Niobium, rod, 500mm, diameter 2.0mm, annealed, 99.9%,
GF08188746 Niobium, tube, 1000mm, outside diameter 5.0mm, inside diameter 4.4mm, wall thickness 0.3mm, as drawn, 99.9%,
GF08129707 Niobium, tube, 200mm, outside diameter 1.0mm, inside diameter 0.5mm, wall thickness 0.25mm, as drawn, 99.9%,
GF37143255 Niobium, tube, 500mm, outside diameter 10.0mm, inside diameter 9.2mm, wall thickness 0.4mm, as drawn, 99.9%,
GF15099778 Niobium, tube, 100mm, outside diameter 10.0mm, inside diameter 9.2mm, wall thickness 0.4mm, as drawn, 99.9%,
GF78772281 Niobium, tube, 100mm, outside diameter 16.0mm, inside diameter 13mm, wall thickness 1.5mm, as drawn, 99.9%,
GF99210630 Niobium, tube, 100mm, outside diameter 2.54mm, inside diameter 2.44mm, wall thickness 0.05mm, annealed, 99.9%,
GF26229359 Niobium, tube, 100mm, outside diameter 5.0mm, inside diameter 4.4mm, wall thickness 0.3mm, as drawn, 99.9%,
GF65944716 Niobium, tube, 200mm, outside diameter 10.0mm, inside diameter 9.2mm, wall thickness 0.4mm, as drawn, 99.9%,
GF27121160 Niobium, tube, 200mm, outside diameter 16.0mm, inside diameter 13mm, wall thickness 1.5mm, as drawn, 99.9%,
GF30503247 Niobium, tube, 200mm, outside diameter 2.54mm, inside diameter 2.44mm, wall thickness 0.05mm, annealed, 99.9%,
GF78669321 Niobium, tube, 200mm, outside diameter 5.0mm, inside diameter 4.4mm, wall thickness 0.3mm, as drawn, 99.9%,
GF16212877 Niobium, tube, 500mm, outside diameter 1.0mm, inside diameter 0.5mm, wall thickness 0.25mm, as drawn, 99.9%,
GF75458245 Niobium, tube, 500mm, outside diameter 16.0mm, inside diameter 13mm, wall thickness 1.5mm, as drawn, 99.9%,
GF28799667 Niobium, tube, 500mm, outside diameter 2.54mm, inside diameter 2.44mm, wall thickness 0.05mm, annealed, 99.9%,
GF33665508 Niobium, tube, 500mm, outside diameter 5.0mm, inside diameter 4.4mm, wall thickness 0.3mm, as drawn, 99.9%,
GF26989587 Niobium, wire reel, 0.5m, diameter 0.18mm, annealed, 99.9%,
GF33194909 Niobium, wire reel, 0.5m, diameter 0.25mm, annealed, 99.9%,
GF91075462 Niobium, wire reel, 0.5m, diameter 0.5mm, annealed, 99.9%,
GF70988013 Niobium, wire reel, 0.5m, diameter 1.0mm, annealed, 99.9%,
GF61174852 Niobium, wire reel, 0.5m, diameter 1.5mm, annealed, 99.9%,
GF45498831 Niobium, wire reel, 100m, diameter 0.18mm, annealed, 99.9%,
GF69435692 Niobium, wire reel, 100m, diameter 0.25mm, annealed, 99.9%,
GF61617348 Niobium, wire reel, 100m, diameter 0.5mm, annealed, 99.9%,
GF88738048 Niobium, wire reel, 10m, diameter 0.05mm, as drawn, 99.85%,
GF46474308 Niobium, wire reel, 10m, diameter 0.125mm, annealed, 99.9%,
GF42178800 Niobium, wire reel, 10m, diameter 1.0mm, annealed, 99.9%,
GF77763055 Niobium, wire reel, 10m, diameter 1.5mm, annealed, 99.9%,
GF21465288 Niobium, wire reel, 1m, diameter 0.05mm, as drawn, 99.85%,
GF15721130 Niobium, wire reel, 1m, diameter 0.18mm, annealed, 99.9%,
GF49363443 Niobium, wire reel, 1m, diameter 0.25mm, annealed, 99.9%,
GF84433642 Niobium, wire reel, 1m, diameter 0.5mm, annealed, 99.9%,
GF73501859 Niobium, wire reel, 1m, diameter 1.0mm, annealed, 99.9%,
GF46856290 Niobium, wire reel, 1m, diameter 1.5mm, annealed, 99.9%,
GF79461939 Niobium, wire reel, 200m, diameter 0.25mm, annealed, 99.9%,
GF72350579 Niobium, wire reel, 20m, diameter 1.0mm, annealed, 99.9%,
GF20382421 Niobium, wire reel, 20m, diameter 1.5mm, annealed, 99.9%,
GF19302768 Niobium, wire reel, 25m, diameter 0.125mm, annealed, 99.9%,
GF42532961 Niobium, wire reel, 25m, diameter 0.18mm, annealed, 99.9%,
GF03755482 Niobium, wire reel, 25m, diameter 0.25mm, annealed, 99.9%,
GF75639145 Niobium, wire reel, 25m, diameter 0.5mm, annealed, 99.9%,
GF83160873 Niobium, wire reel, 2m, diameter 0.05mm, as drawn, 99.85%,
GF43822100 Niobium, wire reel, 2m, diameter 1.5mm, annealed, 99.9%,
GF61065558 Niobium, wire reel, 50m, diameter 0.125mm, annealed, 99.9%,
GF54892930 Niobium, wire reel, 50m, diameter 0.5mm, annealed, 99.9%,
GF95109903 Niobium, wire reel, 5m, diameter 0.05mm, as drawn, 99.85%,
GF02771793 Niobium, wire reel, 5m, diameter 0.125mm, annealed, 99.9%,
GF15199892 Niobium, wire reel, 5m, diameter 0.18mm, annealed, 99.9%,
GF70430048 Niobium, wire reel, 5m, diameter 0.25mm, annealed, 99.9%,
GF54205118 Niobium, wire reel, 5m, diameter 0.5mm, annealed, 99.9%,
GF87731545 Niobium, wire reel, 5m, diameter 1.0mm, annealed, 99.9%,
GF57036304 Niobium, wire reel, 5m, diameter 1.5mm, annealed, 99.9%,
GF59419503 Niobium, insulated wire, 10m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyimide insulation,
GF04567057 Niobium, insulated wire, 10m, conductor diameter 0.05mm, insulation thickness 0.008mm, PTFE (polytetrafluoroethylene) insulation,
GF52723638 Niobium, insulated wire, 10m, conductor diameter 0.125mm, insulation thickness 0.01mm, polyimide insulation,
GF60162331 Niobium, insulated wire, 1m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyimide insulation,
GF72781162 Niobium, insulated wire, 1m, conductor diameter 0.05mm, insulation thickness 0.008mm, PTFE (polytetrafluoroethylene) insulation,
GF41360868 Niobium, insulated wire, 1m, conductor diameter 0.125mm, insulation thickness 0.01mm, polyimide insulation,
GF27438206 Niobium, insulated wire, 20m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyimide insulation,
GF00225447 Niobium, foil, thickness 0.25 mm, 4 mm diameter, purity 99.9%,
GF00481586 Niobium, foil, thickness 0.05 mm, 6 mm diameter, purity 99.9%,
GF00490407 Niobium, foil, thickness 0.05 mm, 25 mm diameter, purity 99.9%,
GF00690780 Niobium, foil, thickness 0.002 mm, size 50 × 50 mm, purity 99.9%,
GF00790107 Niobium, foil, thickness 0.007 mm, size 100 × 100 mm, purity 99.9%,
GF01828694 Niobium, foil, thickness 2.0 mm, size 100 × 100 mm, purity 99.9%,
GF02227597 Niobium, foil, thickness 0.05 mm, size 25 × 25 mm, purity 99.9%,
GF02255947 Niobium, foil, thickness 0.0075 mm, size 25 × 25 mm, purity 99.9%,
GF02569237 Niobium, foil, thickness 0.025 mm, length 1 m, purity 99.9%,
GF02882516 Niobium, foil, thickness 0.005 mm, size 100 × 100 mm, purity 99.9%,
GF02917920 Niobium, foil, thickness 0.0125 mm, 15 mm diameter, purity 99.9%,
GF03576742 Niobium, foil, thickness 0.008 mm, size 50 × 50 mm, purity 99.9%,
GF04466808 Niobium, foil, thickness 0.125 mm, size 300 × 300 mm, purity 99.9%,
GF04603047 Niobium, wire, 1.25 mm diameter, length 10 m, purity 99.9%,
GF04920915 Niobium, nut, size m5, length 25 pcs, style hexagonal,
GF05256592 Niobium, foil, thickness 0.0125 mm, 4 mm diameter, purity 99.9%,
GF05873283 Niobium, foil, thickness 0.30 mm, 6 mm diameter, purity 99.9%,
GF05896435 Niobium, foil, thickness 0.25 mm, size 150 × 150 mm, purity 99.9%,
GF06376079 Niobium, foil, thickness 0.007 mm, 10 mm diameter, purity 99.9%,
GF06435840 Niobium, Washer, size m4, length 50 pcs, outside diameter 8.64 - 9.00mm,
GF06454830 Niobium, foil, thickness 0.003 mm, size 150 × 150 mm, purity 99.9%,
GF07389805 Niobium, Washer, size m3, length 25 pcs, outside diameter 6.64 - 7.00mm,
GF07938513 Niobium, foil, thickness 1.5 mm, size 150 × 150 mm, purity 99.9%,
GF08259486 Niobium, foil, thickness 0.004 mm, 8 mm diameter, purity 99.9%,
GF08836432 Niobium, foil, thickness 0.007 mm, size 50 × 50 mm, purity 99.9%,
GF08920885 Niobium, foil, thickness 0.004 mm, 25 mm diameter, purity 99.9%,
GF09114892 Niobium, foil, thickness 0.02 mm, 15 mm diameter, purity 99.9%,
GF09713493 Niobium, foil, thickness 0.025 mm, size 100 × 100 mm, purity 99.9%,
GF10378100 Niobium, foil, thickness 0.025 mm, 8 mm diameter, purity 99.9%,
GF10680353 Niobium, foil, thickness 0.25 mm, size 50 × 50 mm, purity 99.9%,
GF11589712 Niobium, foil, thickness 0.006 mm, size 150 × 150 mm, purity 99.9%,
GF12152539 Niobium, foil, thickness 0.003 mm, size 25 × 25 mm, purity 99.9%,
GF12220142 Niobium, foil, thickness 0.125 mm, size 25 × 25 mm, purity 99.9%,
GF12376942 Niobium, foil, thickness 0.05 mm, 4 mm diameter, purity 99.9%,
GF12563917 Niobium, Washer, size m4, length 25 pcs, outside diameter 8.64 - 9.00mm,
GF12732016 Niobium, foil, thickness 0.025 mm, size 25 × 25 mm, purity 99.9%,
GF12920992 Niobium, foil, thickness 0.004 mm, size 50 × 50 mm, purity 99.9%,
GF12949319 Niobium, foil, thickness 0.01 mm, size 214 × 300 mm, purity 99.9%,
GF13256535 Niobium, foil, thickness 0.008 mm, size 100 × 100 mm, purity 99.9%,
GF13297697 Niobium, nut, size m4, length 50 pcs, style hexagonal,
GF13380757 Niobium, foil, thickness 0.0125 mm, size 150 × 150 mm, purity 99.9%,
GF13559126 Niobium, foil, thickness 0.006 mm, size 50 × 50 mm, purity 99.9%,
GF13596891 Niobium, foil, thickness 0.15 mm, 50 mm diameter, purity 99.9%,
GF14100860 Niobium, foil, thickness 0.05 mm, 15 mm diameter, purity 99.9%,
GF14604667 Niobium, foil, thickness 0.0075 mm, 4 mm diameter, purity 99.9%,
GF14870987 Niobium, Bolt, size m5 x 30 mm, length 50 pcs, head style hexagonal,
GF16442669 Niobium, foil, thickness 0.25 mm, 10 mm diameter, purity 99.9%,
GF16538137 Niobium, foil, thickness 0.05 mm, length 0.5 m, purity 99.9%,
GF16769098 Niobium, foil, thickness 0.007 mm, size 100 × 100 mm, purity 99.9%,
GF16793129 Niobium, foil, thickness 0.5 mm, 10 mm diameter, purity 99.9%,
GF17036917 Niobium, nut, size m4, length 25 pcs, style hexagonal,
GF17260264 Niobium, foil, thickness 0.1 mm, size 50 × 50 mm, purity 99.9%,
GF17432412 Niobium, foil, thickness 0.25 mm, 6 mm diameter, purity 99.9%,
GF18680474 Niobium, foil, thickness 0.025 mm, size 25 × 25 mm, purity 99.9%,
GF18750810 Niobium, wire, 1.25 mm diameter, length 0.5 m, purity 99.9%,
GF18962501 Niobium, foil, thickness 0.1 mm, 6 mm diameter, purity 99.9%,
GF19413873 Niobium, insulated wire, conductor diameter 0.10 mm, length 2 m, insulation thickness 0.01 mm,
GF19608475 Niobium, wire, 1.25 mm diameter, length 1 m, purity 99.9%,
GF19632876 Niobium, Bolt, size m4 x 30 mm, length 25 pcs, head style hexagonal,
GF20425227 Niobium, foil, thickness 0.02 mm, size 50 × 50 mm, purity 99.9%,
GF20688554 Niobium, foil, thickness 0.30 mm, 25 mm diameter, purity 99.9%,
GF20858205 Niobium, Washer, size m6, length 50 pcs, outside diameter 11.57 - 12.00mm,
GF20883292 Niobium, foil, thickness 0.25 mm, 50 mm diameter, purity 99.9%,
GF21084779 Niobium, foil, thickness 0.008 mm, 8 mm diameter, purity 99.9%,
GF21511291 Niobium, foil, thickness 0.009 mm, size 100 × 100 mm, purity 99.9%,
GF21729551 Niobium, foil, thickness 0.15 mm, size 50 × 50 mm, purity 99.9%,
GF21839182 Niobium, foil, thickness 0.25 mm, size 100 × 100 mm, purity 99.9%,
GF22042044 Niobium, foil, thickness 0.15 mm, 15 mm diameter, purity 99.9%,
GF22345309 Niobium, foil, thickness 1.0 mm, size 100 × 100 mm, purity 99.9%,
GF22619561 Niobium, foil, thickness 0.025 mm, size 50 × 50 mm, purity 99.9%,
GF22634085 Niobium, foil, thickness 0.25 mm, 8 mm diameter, purity 99.9%,
GF23158582 Niobium, foil, thickness 1.5 mm, size 25 × 25 mm, purity 99.9%,
GF23685790 Niobium, foil, thickness 0.006 mm, 25 mm diameter, purity 99.9%,
GF24315117 Niobium, foil, thickness 0.006 mm, 10 mm diameter, purity 99.9%,
GF24667757 Niobium, foil, thickness 0.05 mm, length 0.2 m, purity 99.9%,
GF24712972 Niobium, foil, thickness 0.05 mm, 10 mm diameter, purity 99.9%,
GF24871529 Niobium, foil, thickness 0.006 mm, size 50 × 50 mm, purity 99.9%,
GF24926452 Niobium, nut, size m6, length 50 pcs, style hexagonal,
GF25582765 Niobium, foil, thickness 0.0075 mm, 6 mm diameter, purity 99.9%,
GF26171636 Niobium, foil, thickness 0.007 mm, 25 mm diameter, purity 99.9%,
GF26785435 Niobium, foil, thickness 1.0 mm, size 25 × 25 mm, purity 99.9%,
GF26850191 Niobium, foil, thickness 0.15 mm, size 150 × 150 mm, purity 99.9%,
GF26950619 Niobium, foil, thickness 0.30 mm, size 150 × 150 mm, purity 99.9%,
GF27517855 Niobium, foil, thickness 0.125 mm, size 50 × 50 mm, purity 99.9%,
GF28034502 Niobium, foil, thickness 0.02 mm, size 100 × 100 mm, purity 99.9%,
GF28181077 Niobium, foil, thickness 0.25 mm, 6 mm diameter, purity 99.9%,
GF28270357 Niobium, foil, thickness 0.15 mm, 8 mm diameter, purity 99.9%,
GF28361741 Niobium, foil, thickness 0.125 mm, 4 mm diameter, purity 99.9%,
GF28838021 Niobium, foil, thickness 0.02 mm, size 100 × 100 mm, purity 99.9%,
GF29260784 Niobium, foil, thickness 0.25 mm, 10 mm diameter, purity 99.9%,
GF29898365 Niobium, foil, thickness 0.25 mm, size 150 × 150 mm, purity 99.9%,
GF30679421 Niobium, foil, thickness 0.0075 mm, 10 mm diameter, purity 99.9%,
GF30769263 Niobium, foil, thickness 0.01 mm, size 50 × 50 mm, purity 99.9%,
GF30881550 Niobium, foil, thickness 0.006 mm, 6 mm diameter, purity 99.9%,
GF31520258 Niobium, Bolt, size m3 x 30 mm, length 25 pcs, head style hexagonal,
GF31571388 Niobium, foil, thickness 0.005 mm, size 25 × 25 mm, purity 99.9%,
GF31857835 Niobium, foil, thickness 0.001 mm, size 25 × 25 mm, purity 99.9%,
GF32435771 Niobium, foil, thickness 0.004 mm, 10 mm diameter, purity 99.9%,
GF32602991 Niobium, foil, thickness 0.25 mm, size 25 × 25 mm, purity 99.9%,
GF33164580 Niobium, Bolt, size m6 x 30 mm, length 25 pcs, head style hexagonal,
GF33240327 Niobium, foil, thickness 0.1 mm, 15 mm diameter, purity 99.9%,
GF33385664 Niobium, foil, thickness 0.01 mm, 50 mm diameter, purity 99.9%,
GF34160756 Niobium, foil, thickness 0.30 mm, 8 mm diameter, purity 99.9%,
GF34259384 Niobium, foil, thickness 0.0125 mm, 8 mm diameter, purity 99.9%,
GF34294046 Niobium, foil, thickness 0.5 mm, size 50 × 50 mm, purity 99.9%,
GF34301809 Niobium, foil, thickness 0.30 mm, length 0.2 m, purity 99.9%,
GF34342927 Niobium, foil, thickness 0.004 mm, 4 mm diameter, purity 99.9%,
GF34660829 Niobium, foil, thickness 0.125 mm, 50 mm diameter, purity 99.9%,
GF34728341 Niobium, foil, thickness 0.05 mm, 8 mm diameter, purity 99.9%,
GF34928118 Niobium, foil, thickness 0.0075 mm, 15 mm diameter, purity 99.9%,
GF35059361 Niobium, foil, thickness 0.025 mm, length 0.2 m, purity 99.9%,
GF35181884 Niobium, foil, thickness 0.003 mm, 25 mm diameter, purity 99.9%,
GF35283023 Niobium, foil, thickness 0.025 mm, 6 mm diameter, purity 99.9%,
GF35428653 Niobium, insulated wire, conductor diameter 0.250 mm, length 1 m, insulation thickness 0.04 mm,
GF35596378 Niobium, foil, thickness 0.008 mm, 10 mm diameter, purity 99.9%,
GF35679819 Niobium, foil, thickness 0.125 mm, 10 mm diameter, purity 99.9%,
GF35918602 Niobium, foil, thickness 0.009 mm, 25 mm diameter, purity 99.9%,
GF37218685 Niobium, foil, thickness 0.003 mm, 6 mm diameter, purity 99.9%,
GF37295714 Niobium, foil, thickness 0.25 mm, size 300 × 300 mm, purity 99.9%,
GF38979499 Niobium, nut, size m3, length 25 pcs, style hexagonal,
GF39362025 Niobium, foil, thickness 0.02 mm, 25 mm diameter, purity 99.9%,
GF39389484 Niobium, foil, thickness 0.006 mm, 15 mm diameter, purity 99.9%,
GF39623755 Niobium, foil, thickness 0.0025 mm, size 100 × 100 mm, purity 99.9%,
GF39633655 Niobium, Bolt, size m3 x 30 mm, length 50 pcs, head style hexagonal,
GF39670331 Niobium, foil, thickness 0.05 mm, size 25 × 25 mm, purity 99.9%,
GF39731485 Niobium, foil, thickness 0.125 mm, size 100 × 100 mm, purity 99.9%,
GF40263241 Niobium, foil, thickness 0.05 mm, 25 mm diameter, purity 99.9%,
GF40562917 Niobium, foil, thickness 0.003 mm, size 100 × 100 mm, purity 99.9%,
GF41005718 Niobium, powder, max. particle size 74 micron, weight 200 g, purity 99.85%,
GF41260913 Niobium, foil, thickness 2.0 mm, size 25 × 25 mm, purity 99.9%,
GF41656732 Niobium, foil, thickness 0.15 mm, 25 mm diameter, purity 99.9%,
GF41960651 Niobium, foil, thickness 0.003 mm, 10 mm diameter, purity 99.9%,
GF42198048 Niobium, foil, thickness 0.0075 mm, size 200 × 300 mm, purity 99.9%,
GF42976963 Niobium, foil, thickness 0.0075 mm, size 100 × 100 mm, purity 99.9%,
GF43251005 Niobium, foil, thickness 0.007 mm, 8 mm diameter, purity 99.9%,
GF44036287 Niobium, insulated wire, conductor diameter 0.10 mm, length 1 m, insulation thickness 0.01 mm,
GF44282268 Niobium, foil, thickness 0.025 mm, 4 mm diameter, purity 99.9%,
GF44284147 Niobium, foil, thickness 0.004 mm, size 25 × 25 mm, purity 99.9%,
GF44297747 Niobium, foil, thickness 0.003 mm, 4 mm diameter, purity 99.9%,
GF44388805 Niobium, foil, thickness 0.5 mm, size 150 × 150 mm, purity 99.9%,
GF44475813 Niobium, foil, thickness 0.025 mm, 25 mm diameter, purity 99.9%,
GF44938086 Niobium, foil, thickness 0.1 mm, size 25 × 25 mm, purity 99.9%,
GF45081457 Niobium, foil, thickness 0.003 mm, 15 mm diameter, purity 99.9%,
GF45218280 Niobium, foil, thickness 0.125 mm, 6 mm diameter, purity 99.9%,
GF45601804 Niobium, foil, thickness 0.25 mm, length 0.5 m, purity 99.9%,
GF46024354 Niobium, foil, thickness 0.05 mm, size 150 × 150 mm, purity 99.9%,
GF46157621 Niobium, foil, thickness 0.01 mm, size 100 × 100 mm, purity 99.9%,
GF46423065 Niobium, foil, thickness 0.005 mm, size 50 × 50 mm, purity 99.9%,
GF47579327 Niobium, foil, thickness 0.009 mm, size 150 × 150 mm, purity 99.9%,
GF47813441 Niobium, foil, thickness 0.009 mm, 8 mm diameter, purity 99.9%,
GF47894417 Niobium, foil, thickness 0.02 mm, 4 mm diameter, purity 99.9%,
GF48116976 Niobium, foil, thickness 0.0075 mm, size 50 × 50 mm, purity 99.9%,
GF48134347 Niobium, foil, thickness 0.5 mm, 8 mm diameter, purity 99.9%,
GF48178761 Niobium, foil, thickness 1.0 mm, size 50 × 50 mm, purity 99.9%,
GF48348692 Niobium, foil, thickness 0.007 mm, size 150 × 150 mm, purity 99.9%,
GF48530044 Niobium, foil, thickness 0.0025 mm, size 25 × 25 mm, purity 99.9%,
GF49547290 Niobium, foil, thickness 0.008 mm, 15 mm diameter, purity 99.9%,
GF50195785 Niobium, foil, thickness 0.02 mm, 8 mm diameter, purity 99.9%,
GF50782507 Niobium, foil, thickness 0.009 mm, 6 mm diameter, purity 99.9%,
GF51034452 Niobium, foil, thickness 0.0125 mm, size 50 × 50 mm, purity 99.9%,
GF51743662 Niobium, foil, thickness 0.1 mm, length 0.2 m, purity 99.9%,
GF51797166 Niobium, foil, thickness 0.0075 mm, 50 mm diameter, purity 99.9%,
GF52285439 Niobium, foil, thickness 0.125 mm, 8 mm diameter, purity 99.9%,
GF52335682 Niobium, foil, thickness 0.003 mm, 8 mm diameter, purity 99.9%,
GF52671528 Niobium, wire, 1.25 mm diameter, length 5 m, purity 99.9%,
GF52863509 Niobium, foil, thickness 1.5 mm, size 100 × 100 mm, purity 99.9%,
GF53621951 Niobium, foil, thickness 0.025 mm, size 25 × 25 mm, purity 99.9%,
GF54471360 Niobium, foil, thickness 0.004 mm, 15 mm diameter, purity 99.9%,
GF54847255 Niobium, powder, max. particle size 74 micron, weight 50 g, purity 99.85%,
GF55040789 Niobium, foil, thickness 0.1 mm, 25 mm diameter, purity 99.9%,
GF55367433 Niobium, foil, thickness 0.02 mm, size 150 × 150 mm, purity 99.9%,
GF55576301 Niobium, insulated wire, conductor diameter 0.10 mm, length 10 m, insulation thickness 0.01 mm,
GF56539019 Niobium, foil, thickness 0.25 mm, 25 mm diameter, purity 99.9%,
GF56691210 Niobium, foil, thickness 0.006 mm, size 100 × 100 mm, purity 99.9%,
GF57074070 Niobium, foil, thickness 0.0125 mm, 6 mm diameter, purity 99.9%,
GF57532912 Niobium, foil, thickness 0.1 mm, size 100 × 100 mm, purity 99.9%,
GF57598352 Niobium, foil, thickness 0.25 mm, 15 mm diameter, purity 99.9%,
GF57614621 Niobium, foil, thickness 2.0 mm, size 50 × 50 mm, purity 99.9%,
GF57722429 Niobium, Washer, size m6, length 25 pcs, outside diameter 11.57 - 12.00mm,
GF57955809 Niobium, foil, thickness 0.025 mm, 15 mm diameter, purity 99.9%,
GF58350911 Niobium, foil, thickness 0.125 mm, length 0.2 m, purity 99.9%,
GF58674358 Niobium, foil, thickness 0.5 mm, size 25 × 25 mm, purity 99.9%,
GF59476540 Niobium, foil, thickness 0.025 mm, 50 mm diameter, purity 99.9%,
GF59484561 Niobium, foil, thickness 0.009 mm, size 25 × 25 mm, purity 99.9%,
GF59528043 Niobium, powder, 74 max. part. size (micron), weight 100 g, purity 99.85%,
GF59976613 Niobium, foil, thickness 0.125 mm, diameter 15 mm, purity 99.9%,
GF60194527 Niobium, foil, thickness 0.006 mm, diameter 8 mm, purity 99.9%,
GF60197700 Niobium, foil, thickness 0.05 mm, diameter 8 mm, purity 99.9%,
GF60494047 Niobium, foil, thickness 0.004 mm, purity 99.9%, size 25 × 25 mm,
GF60731603 Niobium, foil, thickness 0.15 mm, size 25 × 25 mm, purity 99.9%,
GF60870258 Niobium, nut, size m3, length 50 pcs, style hexagonal,
GF61765500 Niobium, foil, thickness 0.30 mm, diameter 15 mm, purity 99.9%,
GF61964815 Niobium, foil, thickness 0.25 mm, length 0.2 m, purity 99.9%,
GF62105903 Niobium, foil, size 25 × 25 mm, thickness 0.008 mm, purity 99.9%,
GF62203878 Niobium, foil, thickness 0.006 mm, size 25 × 25 mm, purity 99.9%,
GF63888799 Niobium, bolt, length 25 pcs, size m5 x 30 mm, head style hexagonal,
GF64740146 Niobium, foil, thickness 0.0125 mm, diameter 25 mm, purity 99.9%,
GF65589339 Niobium, foil, diameter 15 mm, thickness 0.01 mm, purity 99.9%,
GF65646984 Niobium, foil, thickness 0.30 mm, diameter 4 mm, purity 99.9%,
GF66014377 Niobium, foil, thickness 0.0075 mm, diameter 25 mm, purity 99.9%,
GF66435734 Niobium, foil, thickness 0.125 mm, size 150 × 150 mm, purity 99.9%,
GF66846370 Niobium, foil, thickness 0.05 mm, size 100 × 100 mm, purity 99.9%,
GF66936651 Niobium, foil, thickness 0.008 mm, size 150 × 150 mm, purity 99.9%,
GF67368673 Niobium, foil, thickness 0.008 mm, diameter 6 mm, purity 99.9%,
GF67958511 Niobium, foil, thickness 0.1 mm, length 0.5 m, purity 99.9%,
GF69104471 Niobium, foil, size 50 × 50 mm, thickness 0.05 mm, purity 99.9%,
GF69703599 Niobium, foil, thickness 0.01 mm, size 150 × 150 mm, purity 99.9%,
GF70094123 Niobium, foil, thickness 0.30 mm, diameter 10 mm, purity 99.9%,
GF70312520 Niobium, foil, thickness 0.1 mm, diameter 8 mm, purity 99.9%,
GF70470570 Niobium, insulated wire, conductor diameter 0.10 mm, length 5 m, insulation thickness 0.01 mm,
GF70587119 Niobium, lump, 5 mm max. lump size, weight 100 g, purity 99.9%,
GF70647059 Niobium, foil, thickness 0.006 mm, diameter 4 mm, purity 99.9%,
GF70746004 Niobium, foil, thickness 0.007 mm, diameter 6 mm, purity 99.9%,
GF70924710 Niobium, foil, thickness 0.01 mm, diameter 10 mm, purity 99.9%,
GF71045423 Niobium, foil, thickness 0.0075 mm, size 150 × 150 mm, purity 99.9%,
GF72151703 Niobium, foil, thickness 0.1 mm, diameter 4 mm, purity 99.9%,
GF72405895 Niobium, foil, thickness 0.001 mm, size 50 × 50 mm, purity 99.9%,
GF72757794 Niobium, lump, 5 mm max. lump size, weight 50 g, purity 99.9%,
GF72935017 Niobium, foil, thickness 0.0025 mm, size 25 × 25 mm, purity 99.9%,
GF73208279 Niobium, foil, thickness 0.0125 mm, size 25 × 25 mm, purity 99.9%,
GF74120059 Niobium, foil, thickness 0.002 mm, size 25 × 25 mm, purity 99.9%,
GF74167397 Niobium, foil, thickness 0.01 mm, diameter 8 mm, purity 99.9%,
GF74335732 Niobium, foil, thickness 0.25 mm, diameter 50 mm, purity 99.9%,
GF74472914 Niobium, foil, thickness 0.02 mm, diameter 10 mm, purity 99.9%,
GF74646153 Niobium, foil, thickness 0.01 mm, size 25 × 25 mm, purity 99.9%,
GF74993338 Niobium, foil, thickness 0.01 mm, diameter 6 mm, purity 99.9%,
GF75023883 Niobium, foil, thickness 0.5 mm, diameter 4 mm, purity 99.9%,
GF75374714 Niobium, foil, thickness 0.25 mm, size 50 × 50 mm, purity 99.9%,
GF75392567 Niobium, foil, thickness 0.004 mm, size 150 × 150 mm, purity 99.9%,
GF75876418 Niobium, lump, 5 mm max. lump size, weight 200 g, purity 99.9%,
GF76231199 Niobium, foil, thickness 0.05 mm, length 1 m, purity 99.9%,
GF76680692 Niobium, foil, thickness 0.009 mm, diameter 10 mm, purity 99.9%,
GF76789120 Niobium, foil, thickness 0.30 mm, size 300 × 300 mm, purity 99.9%,
GF76967869 Niobium, washer, size m3, length 50 pcs, outside diameter 6.64 - 7.00mm,
GF77985073 Niobium, foil, thickness 0.5 mm, diameter 6 mm, purity 99.9%,
GF78449024 Niobium, foil, thickness 0.05 mm, size 50 × 50 mm, purity 99.9%,
GF78779794 Niobium, foil, thickness 0.30 mm, size 100 × 100 mm, purity 99.9%,
GF79016205 Niobium, wire, 1.25 mm diameter, purity 99.9%, length 20 m,
GF79092029 Niobium, foil, thickness 0.02 mm, size 25 × 25 mm, purity 99.9%,
GF79307040 Niobium, foil, thickness 0.0125 mm, size 100 × 100 mm, purity 99.9%,
GF79706795 Niobium, foil, thickness 0.15 mm, length 0.2 m, purity 99.9%,
GF79775623 Niobium, foil, thickness 0.25 mm, diameter 15 mm, purity 99.9%,
GF79872047 Niobium, foil, thickness 0.25 mm, size 25 × 25 mm, purity 99.9%,
GF80097767 Niobium, foil, thickness 0.1 mm, diameter 50 mm, purity 99.9%,
GF80388018 Niobium, foil, thickness 0.009 mm, size 50 × 50 mm, purity 99.9%,
GF81069835 Niobium, foil, thickness 0.008 mm, size 100 × 100 mm, purity 99.9%,
GF81199053 Niobium, nut, size m6, length 25 pcs, style hexagonal,
GF81277106 Niobium, washer, size m5, length 50 pcs, outside diameter 9.64 - 10.00mm,
GF81499371 Niobium, foil, thickness 0.05 mm, diameter 6 mm, purity 99.9%,
GF81692208 Niobium, foil, thickness 0.5 mm, diameter 50 mm, purity 99.9%,
GF82124850 Niobium, foil, thickness 0.007 mm, size 25 × 25 mm, purity 99.9%,
GF82403850 Niobium, foil, thickness 0.05 mm, diameter 4 mm, purity 99.9%,
GF82420556 Niobium, foil, thickness 0.025 mm, length 0.5 m, purity 99.9%,
GF82423346 Niobium, foil, thickness 0.0125 mm, diameter 10 mm, purity 99.9%,
GF82582487 Niobium, nut, size m5, length 50 pcs, style hexagonal,
GF82999940 Niobium, foil, diameter 4 mm, thickness 0.01 mm, purity 99.9%,
GF83382824 Niobium, foil, thickness 0.02 mm, purity 99.9%, size 150 × 150 mm,
GF83492972 Niobium, foil, thickness 0.15 mm, size 100 × 100 mm, purity 99.9%,
GF84647144 Niobium, foil, thickness 0.15 mm, diameter 4 mm, purity 99.9%,
GF84840093 Niobium, foil, thickness 0.003 mm, size 50 × 50 mm, purity 99.9%,
GF85087345 Niobium, foil, thickness 0.009 mm, diameter 15 mm, purity 99.9%,
GF85206269 Niobium, foil, thickness 0.009 mm, diameter 4 mm, purity 99.9%,
GF85423417 Niobium, foil, thickness 0.15 mm, length 0.5 m, purity 99.9%,
GF86069391 Niobium, foil, thickness 0.30 mm, diameter 50 mm, purity 99.9%,
GF86276830 Niobium, foil, thickness 0.05 mm, diameter 50 mm, purity 99.9%,
GF86755113 Niobium, foil, thickness 0.5 mm, size 100 × 100 mm, purity 99.9%,
GF86801564 Niobium, foil, thickness 0.02 mm, diameter 6 mm, purity 99.9%,
GF87725177 Niobium, foil, thickness 0.30 mm, length 0.5 m, purity 99.9%,
GF89007329 Niobium, bolt, size m4 x 30 mm, length 50 pcs, head style hexagonal,
GF89065929 Niobium, foil, thickness 0.1 mm, diameter 10 mm, purity 99.9%,
GF89122103 Niobium, foil, thickness 0.05 mm, diameter 15 mm, purity 99.9%,
GF89513770 Niobium, bolt, size m6 x 30 mm, length 50 pcs, head style hexagonal,
GF90477962 Niobium, washer, length 25 pcs, size m5, outside diameter 9.64 - 10.00mm,
GF91613943 Niobium, foil, thickness 0.1 mm, size 150 × 150 mm, purity 99.9%,
GF91852577 Niobium, foil, thickness 0.05 mm, diameter 10 mm, purity 99.9%,
GF91966641 Niobium, foil, thickness 0.30 mm, size 25 × 25 mm, purity 99.9%,
GF92256172 Niobium, wire, 0.05 mm diameter, length 20 m, purity 99.85%,
GF92275465 Niobium, foil, thickness 0.15 mm, diameter 6 mm, purity 99.9%,
GF92518678 Niobium, foil, diameter 50 mm, thickness 0.0125 mm, purity 99.9%,
GF92653533 Niobium, foil, thickness 0.5 mm, diameter 25 mm, purity 99.9%,
GF92866078 Niobium, foil, thickness 0.004 mm, diameter 6 mm, purity 99.9%,
GF93110206 Niobium, foil, thickness 0.003 mm, size 25 × 25 mm, purity 99.9%,
GF93162720 Niobium, foil, thickness 1.0 mm, purity 99.9%, size 150 × 150 mm,
GF93365961 Niobium, foil, thickness 0.007 mm, diameter 4 mm, purity 99.9%,
GF93412100 Niobium, foil, thickness 0.008 mm, diameter 25 mm, purity 99.9%,
GF94366424 Niobium, foil, thickness 0.009 mm, size 100 × 100 mm, purity 99.9%,
GF94814219 Niobium, foil, thickness 0.1 mm, length 1 m, purity 99.9%,
GF94822904 Niobium, foil, thickness 1.5 mm, size 50 × 50 mm, purity 99.9%,
GF95279215 Niobium, foil, thickness 0.25 mm, diameter 4 mm, purity 99.9%,
GF95830230 Niobium, foil, thickness 0.30 mm, size 50 × 50 mm, purity 99.9%,
GF96378968 Niobium, foil, thickness 0.004 mm, size 100 × 100 mm, purity 99.9%,
GF96538944 Niobium, foil, thickness 0.008 mm, diameter 4 mm, purity 99.9%,
GF96645998 Niobium, foil, size 300 × 300 mm, thickness 0.25 mm, purity 99.9%,
GF96893128 Niobium, foil, thickness 0.15 mm, diameter 10 mm, purity 99.9%,
GF96999282 Niobium, foil, thickness 0.0025 mm, size 50 × 50 mm, purity 99.9%,
GF97009375 Niobium, foil, diameter 15 mm, thickness 0.007 mm, purity 99.9%,
GF97077280 Niobium, foil, thickness 0.125 mm, diameter 25 mm, purity 99.9%,
GF97263647 Niobium, foil, thickness 0.025 mm, diameter 10 mm, purity 99.9%,
GF97439596 Niobium, insulated wire, conductor diameter 0.10 mm, length 20 m, insulation thickness 0.01 mm,
GF97677703 Niobium, foil, thickness 0.025 mm, size 50 × 50 mm, purity 99.9%,
GF97845430 Niobium, foil, thickness 0.0075 mm, diameter 8 mm, purity 99.9%,
GF97857972 Niobium, foil, thickness 0.25 mm, diameter 8 mm, purity 99.9%,
GF98187081 Niobium, foil, thickness 0.025 mm, size 50 × 50 mm, purity 99.9%,
GF98667084 Niobium, foil, thickness 0.01 mm, diameter 25 mm, purity 99.9%,
GF98934744 Niobium, foil, thickness 0.25 mm, size 100 × 100 mm, purity 99.9%,
GF99141228 Niobium, foil, thickness 0.5 mm, diameter 15 mm, purity 99.9%,
GF99794682 Niobium, foil, thickness 0.25 mm, diameter 25 mm, purity 99.9%,