• USA Home
  • Search Results

  • niodoacetaminoethyl1naphthylamine5sulfonicacid434253693063911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-(Iodoacetaminoethyl)-1-naphthylamine-5-sulfonic acid

1 Products
Synonym:   1,5-I-AEDANS, 5-[2-(Iodoacetamido)ethylamino]naphthalene-1-sulfonic acid, N-Iodoacetyl-N′-(5-sulfo-1-naphthyl)ethylenediamine, Hudson-Weber-Reagent
Image of <I>N</I>-<WBR>(Iodoacetaminoethyl)<WBR>-<WBR>1-<WBR>naphthylamine-<WBR>5-<WBR>sulfonic acid
CAS Number:
Linear Formula:
 ICH2CONHCH2CH2NHC10H6SO3H
Molecular Weight:
 434.25
 Beilstein Registry Number:
 3004454
 EC Number:
 MDL number:
 PubChem Substance ID:
 eCl@ss
 32035301
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Fluorescence Test
Show More