• USA Home
  • Search Results

  • nisobutoxymethylacrylamide157211666959311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-(Isobutoxymethyl)acrylamide

1 Products
Image of <I>N</I>-<WBR>(Isobutoxymethyl)<WBR>acrylamide
CAS Number:
Linear Formula:
 CH2=CHCONHCH2OCH2CH(CH3)2
Molecular Weight:
 157.21
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More