• USA Home
  • Search Results

  • nisoldipine388416367572911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nisoldipine

1 Products
Synonym:   (±)-Isobutyl methyl 1,4-dihydro-2,6-dimethyl-4-(o-nitrophenyl)-3,5-pyridinedicarboxylate, 1,4-Dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylic acid methyl 2-methylpropyl ester, Sular
Image of Nisoldipine
CAS Number:
Empirical Formula (Hill Notation):
 C20H24N2O6
Molecular Weight:
 388.41
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
N0165 Nisoldipine, ≥98% (HPLC),