• USA Home
  • Search Results

  • nitricacid6301769737211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nitric acid

17 Products
Image of Nitric acid
CAS Number:
Empirical Formula (Hill Notation):
 HNO3
Molecular Weight:
 63.01
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
 eCl@ss:
 38060304
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Suitability
  Quality clarifier
  Miscellaneous Specifications
  Product Line
  Reaction Suitability
  Vapor density vs air
  Application(s)
  Ice Shipping
  Melting Point
  pK-Values
  Vapor pressure
  Purity Grade
  Concentration
  Agency/Method
  Restriction
  Color
  pH Value
  Physical Form
  Special Grade
  Density
Show More

Product #
Description
438073 Nitric acid, ACS reagent, 70%,
225711 Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis,
30709 Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., for determinations with dithizone, ≥65%,
695041 Nitric acid, ACS reagent, ≥90.0%,
84378 Nitric acid, puriss. p.a., 65.0-67.0%,
84380 Nitric acid, puriss. p.a., ≥65% (T),
309079 Nitric acid, red, fuming, HNO3 >90 %,
1.60307 Nitric acid, c(HNO3) = 1 mol/l (1 N) Titripur®,
1.60236 Nitric acid, 0.1 M HNO3 (0.1 N), Titripur®,
1.00630 Nitric acid, c(HNOз) = 10 mol/l (10 N) Titripur®,
1.09966 Nitric acid, for 1000 ml, c(HNO₃) = 1 mol/l (1 N) Titrisol®,
695033 Nitric acid, ACS reagent, 70%,
NX0407 Nitric Acid, Aqueous OmniTrace,
NX0408 Nitric Acid, OmniTrace Ultra,
NX0409 Nitric Acid, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS,
NX0412 Nitric Acid,
84381 Nitric acid, puriss. p.a., packed in coated, shock- and leak-protected glass bottle, ≥65% (T),