• USA Home
  • Search Results

  • nitrilotriaceticacid1911413913911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nitrilotriacetic acid

5 Products
Synonym:   N,N-Bis(carboxymethyl)glycine, NTA, Tris(carboxymethyl)amine
Image of Nitrilotriacetic acid
CAS Number:
Linear Formula:
 N(CH2COOH)3
Molecular Weight:
 191.14
 Beilstein/REAXYS Number:
 1710776
 EC Number:
 MDL number:
 NACRES:
 NA.31
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  UV-Absorption Values
  Application(s)
  Loss
  Assay
Show More