• USA Home
  • Search Results

  • nitriteionstandardsolution6900763200011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nitrite ion standard solution

2 Products
Synonym:   Sodium nitrite solution
Image of Nitrite ion standard solution
CAS Number:
Linear Formula:
 NaNO2
Molecular Weight:
 69.00
 MDL number:
 NACRES:
 NB.61
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Shelf Life
  Special Grade
Show More