• USA Home
  • Search Results

  • nitrofurantoin238166720911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nitrofurantoin

4 Products
Synonym:   N-(5-Nitro-2-furfurylidene)-1-aminohydantoin, Furadoxyl, Nitrofurantoine
Image of Nitrofurantoin
CAS Number:
Empirical Formula (Hill Notation):
 C8H6N4O5
Molecular Weight:
 238.16
 Beilstein/REAXYS Number:
 893207
 EC Number:
 MDL number:
 NACRES:
 NA.85
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Assay
  Anal. Standard Form
  Activity Spectrum
  Special Grade
  Physical Form
  Mode of Action
Show More