• USA Home
  • Search Results

  • nitrogenandoxygenisotopesinnitrate10110775779111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nitrogen and oxygen isotopes in nitrate

1 Products
Synonym:   Potassium nitrate
Image of Nitrogen and oxygen isotopes in nitrate
CAS Number:
Empirical Formula (Hill Notation):
 KNO3
Molecular Weight:
 101.10
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More