• USA Home
  • Search Results

  • nitrotetrazoliumbluechloride8176429883911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nitrotetrazolium Blue chloride

3 Products
Synonym:   2,2′-bis(4-Nitrophenyl)-5,5′-diphenyl-3,3′-(3,3′-dimethoxy-4,4′-diphenylene)ditetrazolium chloride, 3,3′-(3,3′-Dimethoxy-4,4′-biphenylene)bis[2-(4-nitrophenyl)-5-phenyl-2H-tetrazolium chloride], p-Nitro-Blue tetrazolium chloride, p-Nitrotetrazolium blue, NBT, Nitro BT
Image of Nitrotetrazolium Blue chloride
CAS Number:
Linear Formula:
 C40H30N10O6 · 2Cl
Molecular Weight:
 817.64
 Beilstein/REAXYS Number:
 4115923
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Application(s)
  Physical Form
  Assay
Show More