• USA Home
  • Search Results

  • nivalenolsolution312322328220411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nivalenol solution

1 Products
Synonym:   3α,4β,7α, 15-Tetrahydroxy-12,13-epoxytrichothec-9-en-8-one, NIV
Image of Nivalenol solution
CAS Number:
Empirical Formula (Hill Notation):
 C15H20O7
Molecular Weight:
 312.32
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More