• USA Home
  • Search Results

  • nlauroylsarcosinesodiumsalt2933813716611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-Lauroylsarcosine sodium salt

5 Products
Synonym:   N-Dodecanoyl-N-methylglycine sodium salt, Sarkosyl NL, Sodium lauroyl sarcosinate
Image of <I>N</I>-Lauroylsarcosine sodium salt
CAS Number:
Linear Formula:
 CH3(CH2)10CON(CH3)CH2COONa
Molecular Weight:
 293.38
 Beilstein/REAXYS Number:
 5322974
 EC Number:
 MDL number:
 NACRES:
 NA.52
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  UV-Absorption Values
  Product Line
  Molecular weight
  Special Grade
  Solubility
  Physical Form
  ID clarifier
  Assay
Show More