• USA Home
  • Search Results

  • nmethyl13diaminopropane8815629184511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-Methyl-1,3-diaminopropane

1 Products
Synonym:   3-(Methylamino)propylamine, N-Methyl-1,3-propanediamine
Image of <I>N</I>-<WBR>Methyl-<WBR>1,3-<WBR>diaminopropane
CAS Number:
Linear Formula:
 CH3NH(CH2)3NH2
Molecular Weight:
 88.15
 Beilstein/REAXYS Number:
 878143
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More