• USA Home
  • Search Results

  • nmethyl1naphthalenemethylaminehydrochloride207706547313411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-Methyl-1-naphthalenemethylamine hydrochloride

1 Products
Image of <I>N</I>-<WBR>Methyl-<WBR>1-<WBR>naphthalenemethylamine hydrochloride
CAS Number:
Linear Formula:
 C10H7CH2NHCH3 · HCl
Molecular Weight:
 207.70
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More