• USA Home
  • Search Results

  • nmethylcytisine2042748686211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-Methylcytisine

3 Products
Synonym:   Caulophylline
Image of <I>N</I>-Methylcytisine
CAS Number:
Empirical Formula (Hill Notation):
 C12H16N2O
Molecular Weight:
 204.27
 EC Number:
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More