• USA Home
  • Search Results

  • nmethylpropargylamine69113516171811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-Methylpropargylamine

1 Products
Synonym:   3-Methylamino-1-propyne
Image of <I>N</I>-Methylpropargylamine
CAS Number:
Linear Formula:
 HC≡CCH2NHCH3
Molecular Weight:
 69.11
 Beilstein/REAXYS Number:
 1071242
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More