• USA Home
  • Search Results

  • nmethylpyrrolidone991387250411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-Methylpyrrolidone

1 Products
Synonym:   1-Methyl-2-pyrrolidinone, 1-Methyl-2-pyrrolidone, N-Methyl-2-pyrrolidone, NMP
Image of N-Methylpyrrolidone
CAS Number:
Empirical Formula (Hill Notation):
 C5H9NO
Molecular Weight:
 99.13
 Beilstein/REAXYS Number:
 106420
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Special Grade
Show More