• USA Home
  • Search Results

  • nnbissalicylideneethylenediamine268319493911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N,N′-Bis(salicylidene)ethylenediamine

1 Products
Synonym:   2,2′-Ethylenebis(nitrilomethylidene)diphenol, N,N′-Ethylenebis(salicylimine)
Image of <I>N</I>,<I>N</I>′-Bis(salicylidene)ethylenediamine
CAS Number:
Linear Formula:
 (CH2N=CHC6H4OH)2
Molecular Weight:
 268.31
 Beilstein/REAXYS Number:
 535296
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More