• USA Home
  • Search Results

  • nndiethyl21naphthyloxypropanamide271351529999711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N,N-Diethyl-2-(1-naphthyloxy)propanamide

1 Products
Image of <I>N</I>,<I>N</I>-<WBR>Diethyl-<WBR>2-<WBR>(1-<WBR>naphthyloxy)<WBR>propanamide
CAS Number:
Linear Formula:
 C10H7OCH(CH3)CON(CH2CH3)2
Molecular Weight:
 271.35
 Beilstein/REAXYS Number:
 6809160
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More