• USA Home
  • Search Results

  • nndiethylnicotinamide178235926711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N,N-Diethylnicotinamide

1 Products
Synonym:   ‘Nikethamide’, DENC, Nicotinic acid diethylamide
Image of <I>N,N</I>-Diethylnicotinamide
CAS Number:
Empirical Formula (Hill Notation):
 C10H14N2O
Molecular Weight:
 178.23
 Beilstein/REAXYS Number:
 5743
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More