• USA Home
  • Search Results

  • nndiethylpphenylenediaminesulfatesalt26233628363211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N,N-Diethyl-p-phenylenediamine sulfate salt

1 Products
Synonym:   4-Amino-N,N-diethylaniline sulfate salt
Image of <I>N,N</I>-Diethyl-<I>p</I>-phenylenediamine sulfate salt
CAS Number:
Linear Formula:
 (C2H5)2NC6H4NH2 · H2SO4
Molecular Weight:
 262.33
 Beilstein/REAXYS Number:
 4219768
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Melting Point
Show More