• USA Home
  • Search Results

  • nndimethyl13propanediamine1021811133111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N,N′-Dimethyl-1,3-propanediamine

1 Products
Synonym:   1,3-Bis(methylamino)propane, N,N′-Dimethyltrimethylenediamine
Image of <I>N,N′</I>-Dimethyl-1,3-propanediamine
CAS Number:
Linear Formula:
 CH3NHCH2CH2CH2NHCH3
Molecular Weight:
 102.18
 Beilstein/REAXYS Number:
 773668
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More