• USA Home
  • Search Results

  • nndimethylacetamide871212719511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N,N-Dimethylacetamide

14 Products
Synonym:   DMAc
Image of <I>N,N</I>-Dimethylacetamide
CAS Number:
Linear Formula:
 CH3CON(CH3)2
Molecular Weight:
 87.12
 Beilstein/REAXYS Number:
 1737614
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Refractive Index
  Vapor pressure
  UV-Absorption Condition
  Transition temperature
  Solubility
  Potency
  Package quantity
  Melting Point
  Explosion limit
  Density
  Autoignition temp
  Product Line
  Application(s)
  UV-Absorption Values
  pH Value
  Physical Form
  Special Grade
Show More

Product #
Description
271012 N,N-Dimethylacetamide, anhydrous, 99.8%,
270555 N,N-Dimethylacetamide, for HPLC, ≥99.9%,
185884 N,N-Dimethylacetamide, ReagentPlus®, ≥99%,
38840 N,N-Dimethylacetamide, puriss. p.a., ≥99.5% (GC),
1.00399 N,N-Dimethylacetamide, for headspace gas chromatography SupraSolv®,
D137510 N,N-Dimethylacetamide, ReagentPlus®, 99%,
ARK2190 N,N-Dimethylacetamide, PharmaGrade, Manufactured under appropriate GMP controls for pharma and biopharmaceutical production., Ph Eur,
8.03235 N,N-Dimethylacetamide, for synthesis,
72336 N,N-Dimethylacetamide, analytical standard,
17308 N,N-Dimethylacetamide, for peptide synthesis, ≥99.8% (GC),
154806 N,N-Dimethylacetamide, spectrophotometric grade, ≥99%,
RTC000104 N,N-Dimethylacetamide, Pharmaceutical Secondary Standard; Certified Reference Material,
PHR2110 N,N-Dimethylacetamide, Pharmaceutical Secondary Standard; Certified Reference Material,
900634 N,N-Dimethylacetamide, anhydrous, ZerO2®, 99.8%,