• USA Home
  • Search Results

  • nndimethylacrylamide9913268003711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N,N-Dimethylacrylamide

1 Products
Synonym:   2-Propenamide
Image of <I>N,N</I>-Dimethylacrylamide
CAS Number:
Linear Formula:
 CH2=CHCON(CH3)2
Molecular Weight:
 99.13
 Beilstein/REAXYS Number:
 1742219
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More