• USA Home
  • Search Results

  • nndimethyldodecylaminenoxide22940164320511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N,N-Dimethyldodecylamine N-oxide

1 Products
Synonym:   DDAO, LDAO, Lauryldimethylamine N-oxide
Image of <I>N,N</I>-Dimethyldodecylamine <I>N</I>-oxide
CAS Number:
Linear Formula:
 CH3(CH2)11N(O)(CH3)2
Molecular Weight:
 229.40
 Beilstein/REAXYS Number:
 1769927
 EC Number:
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More